2-Chloro-4-(difluoromethoxy)pyridine 98%

BLDPBD238970-100MGEA 57 EUR
BLDPBD238970-100MG BLDPBD238970-5G BLDPBD238970-250MG BLDPBD238970-1G
2-Chloro-4-(difluoromethoxy)pyridine 98%
2-Chloro-4-(difluoromethoxy)pyridine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₆H₄ClF₂NO
MW: 179,55 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD13185848
CAS-nummer: 1206978-15-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR