Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%

BLDPBD2399-500GEA 174 EUR
BLDPBD2399-500G BLDPBD2399-10G BLDPBD2399-1000G BLDPBD2399-25G BLDPBD2399-5G BLDPBD2399-100G
Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%
Methyl-2-amino-3-hydroxypropanoate hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₄H₁₀ClNO₃
MW: 155,578 g/mol
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 5619-04-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR