1-Hexanesulphonic acid sodium salt monohydrate 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD240579-5GEA 6 EUR
BLDPBD240579-5G
1-Hexanesulphonic acid sodium salt monohydrate 98%
1-Hexanesulphonic acid sodium salt monohydrate

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₁₃NaO₃S·1H₂O
MW: 206,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00149549
CAS-nummer: 207300-91-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR