Ethyl-3-bromo-1H-pyrazole-4-carboxylate 97%

BLDPBD241844-25GEA 436 EUR
BLDPBD241844-25G BLDPBD241844-1G BLDPBD241844-250MG BLDPBD241844-10G BLDPBD241844-100G BLDPBD241844-100MG BLDPBD241844-5G
Ethyl-3-bromo-1H-pyrazole-4-carboxylate 97%
Ethyl-3-bromo-1H-pyrazole-4-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formule: C₆H₇BrN₂O₂
MW: 219,04 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD12032195
CAS-nummer: 1353100-91-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR