1H-Indazol-7-ol 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD24581-250MGEA 25 EUR
BLDPBD24581-250MG BLDPBD24581-10G BLDPBD24581-25G BLDPBD24581-1G BLDPBD24581-100MG BLDPBD24581-5G
1H-Indazol-7-ol 97%
1H-Indazol-7-ol

Storage condition: Sealed in dry, 2-8°C

Formule: C₇H₆N₂O
MW: 134,1400 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00464298
CAS-nummer: 81382-46-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR