2-Acetyl-6-bromopyridine 98%

BLDPBD25063-250MGEA 6 EUR
BLDPBD25063-250MG BLDPBD25063-500G BLDPBD25063-1G BLDPBD25063-25G BLDPBD25063-5G BLDPBD25063-100G BLDPBD25063-10G
2-Acetyl-6-bromopyridine 98%
2-Acetyl-6-bromopyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₇H₆BrNO
MW: 200,03 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00272200
CAS-nummer: 49669-13-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR