2-Chloro-4-fluoropyridine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD2539-10GEA 24 EUR
BLDPBD2539-10G BLDPBD2539-5G BLDPBD2539-25G BLDPBD2539-1G
2-Chloro-4-fluoropyridine 97%
2-Chloro-4-fluoropyridine

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₃ClFN
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD04039345
CAS-nummer: 34941-91-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR