1-(4-Bromophenylsulphonyl)pyrrolidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD254926-100MGEA 13 EUR
BLDPBD254926-100MG BLDPBD254926-250MG BLDPBD254926-1G
1-(4-Bromophenylsulphonyl)pyrrolidine 98%
1-(4-Bromophenylsulphonyl)pyrrolidine

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₂BrNO₂S
MW: 290,1768 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00584577
CAS-nummer: 136350-52-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR