4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD257297-250MGEA 23 EUR
BLDPBD257297-250MG
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester 98%
4-(3-(Ethoxycarbonyl)piperidine-1-carbonyl)phenylboronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Formule: C₂₁H₃₀BNO₅
MW: 387,28 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD06657751
CAS-nummer: 850411-14-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR