5-Bromo-4-methylpyridine-2-carboxylic acid 98%

BLDPBD26220-1GEA 26 EUR
BLDPBD26220-1G BLDPBD26220-100G BLDPBD26220-5G BLDPBD26220-25G BLDPBD26220-250MG BLDPBD26220-10G
5-Bromo-4-methylpyridine-2-carboxylic acid 98%
5-Bromo-4-methylpyridine-2-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₆BrNO₂
MW: 216,0300 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07375100
CAS-nummer: 886365-02-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR