5-Chloro-3-methylpyridine-2-carboxylic acid 95%

BLDPBD26258-1GEA 23 EUR
BLDPBD26258-1G BLDPBD26258-250MG
5-Chloro-3-methylpyridine-2-carboxylic acid 95%
5-Chloro-3-methylpyridine-2-carboxylic acid

Storage condition: Inert atmosphere, 2-8°C

Formule: C7H6ClNO2
MW: 171,5800 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD07375127
CAS-nummer: 886365-46-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR