Dihydro-2H-pyran-3(4H)-one 95%

BLDPBD26783-5GEA 89 EUR
BLDPBD26783-5G BLDPBD26783-25G BLDPBD26783-1G BLDPBD26783-250MG BLDPBD26783-0.025G BLDPBD26783-10G BLDPBD26783-100G
Dihydro-2H-pyran-3(4H)-one 95%
Dihydro-2H-pyran-3(4H)-one

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₅H₈O₂
MW: 100,1158 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00182426
CAS-nummer: 23462-75-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR