Ethyl-5-formyl-1H-pyrrole-2-carboxylate 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD26829-5GEA 168 EUR
BLDPBD26829-5G BLDPBD26829-250MG BLDPBD26829-10G BLDPBD26829-100MG BLDPBD26829-25G BLDPBD26829-1G
Ethyl-5-formyl-1H-pyrrole-2-carboxylate 97%
Ethyl-5-formyl-1H-pyrrole-2-carboxylate
Formule: C₈H₉NO₃
MW: 167,1600 g/mol
MDL-nummer: MFCD00159462
CAS-nummer: 7126-50-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR