4-Bromo-3,5-difluorobenzonitrile 97%
|
|
BLDPBD272654-250MGEA
47
EUR
BLDPBD272654-250MG
BLDPBD272654-1G
BLDPBD272654-25G
BLDPBD272654-100MG
BLDPBD272654-5G
4-Bromo-3,5-difluorobenzonitrile 97%
4-Bromo-3,5-difluorobenzonitrile
Storage condition: Sealed in dry, 2-8°C
Formule:
C₇H₂BrF₂N MW: 217,9983 g/mol Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD11845964 CAS-nummer: 123688-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...