4-Morpholinobenzaldehyde 97%

BLDPBD27297-25GEA 105 EUR
BLDPBD27297-25G BLDPBD27297-5G BLDPBD27297-10G BLDPBD27297-1G BLDPBD27297-100G
4-Morpholinobenzaldehyde 97%
4-Morpholinobenzaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₁H₁₃NO₂
MW: 191,2264 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00735826
CAS-nummer: 1204-86-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR