1-Boc-4-methylpiperidine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD27320-5GEA 276 EUR
BLDPBD27320-5G BLDPBD27320-1G BLDPBD27320-250MG
1-Boc-4-methylpiperidine 97%
1-Boc-4-methylpiperidine

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₁H₂₁NO₂
MW: 199,2899 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04114947
CAS-nummer: 123387-50-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR