5-Acetyl-2-bromopyridine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD27499-10GEA 63 EUR
BLDPBD27499-10G BLDPBD27499-100G BLDPBD27499-5G BLDPBD27499-250MG BLDPBD27499-25G BLDPBD27499-1G
5-Acetyl-2-bromopyridine 97%
5-Acetyl-2-bromopyridine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₆BrNO
MW: 200,03 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04974527
CAS-nummer: 139042-59-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR