2-Chloropyrimidine-4,5-diamine 97%

BLDPBD27558-25GEA 851 EUR
BLDPBD27558-25G BLDPBD27558-250MG BLDPBD27558-1G BLDPBD27558-100MG BLDPBD27558-5G
2-Chloropyrimidine-4,5-diamine 97%
2-Chloropyrimidine-4,5-diamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₄H₅ClN₄
MW: 144,5623 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00047355
CAS-nummer: 14631-08-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR