Methyl-3-amino-2-chloroisonicotinate 98%

BLDPBD27767-250MGEA 26 EUR
BLDPBD27767-250MG BLDPBD27767-5G BLDPBD27767-25G BLDPBD27767-10G BLDPBD27767-1G
Methyl-3-amino-2-chloroisonicotinate 98%
Methyl-3-amino-2-chloroisonicotinate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₇H₇ClN₂O₂
MW: 186,6000 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03411723
CAS-nummer: 173435-41-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR