tert-Butyl-6-amino-1H-indazole-1-carboxylate 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD28069-250MGEA 17 EUR
BLDPBD28069-250MG BLDPBD28069-1G BLDPBD28069-5G
tert-Butyl-6-amino-1H-indazole-1-carboxylate 97%
tert-Butyl-6-amino-1H-indazole-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₁₂H₁₅N₃O₂
MW: 233,27 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD04114657
CAS-nummer: 219503-81-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR