5-Amino-1-(2-hydroxyethyl)pyrazole 98%

BLDPBD2852-5GEA 24 EUR
BLDPBD2852-5G BLDPBD2852-25G BLDPBD2852-1G BLDPBD2852-0.025G
5-Amino-1-(2-hydroxyethyl)pyrazole 98%
5-Amino-1-(2-hydroxyethyl)pyrazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₅H₉N₃O
MW: 127,15 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08063809
CAS-nummer: 73616-27-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR