3-Acetyl-5-bromopyridine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD28576-250MGEA 6 EUR
BLDPBD28576-250MG BLDPBD28576-10G BLDPBD28576-5G BLDPBD28576-25G BLDPBD28576-1G BLDPBD28576-100G
3-Acetyl-5-bromopyridine 97%
3-Acetyl-5-bromopyridine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₇H₆BrNO
MW: 200,03 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03086033
CAS-nummer: 38940-62-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR