4-(Piperidine-1-carbonyl)phenylboronic acid 98%

BLDPBD28577-250MGEA 31 EUR
BLDPBD28577-250MG BLDPBD28577-5G BLDPBD28577-1G
4-(Piperidine-1-carbonyl)phenylboronic acid 98%
4-(Piperidine-1-carbonyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₂H₁₆BNO₃
MW: 233,0713 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03411951
CAS-nummer: 389621-83-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR