2,5-Difluorobenzamide 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD291135-10GEA 23 EUR
BLDPBD291135-10G BLDPBD291135-5G BLDPBD291135-1G
2,5-Difluorobenzamide 97%
2,5-Difluorobenzamide

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₅F₂NO
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00015548
CAS-nummer: 85118-03-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR