Pyrazine-2-carbaldehyde 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD29122-250MGEA 23 EUR
BLDPBD29122-250MG BLDPBD29122-10G BLDPBD29122-25G BLDPBD29122-0.025G BLDPBD29122-5G BLDPBD29122-1G BLDPBD29122-100MG
Pyrazine-2-carbaldehyde 95%
Pyrazine-2-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formule: C5H4N2O
MW: 108,0981 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD02179278
CAS-nummer: 5780-66-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR