2-Acetyl-4-methylpyridine 98%

BLDPBD29187-1GEA 11 EUR
BLDPBD29187-1G BLDPBD29187-0.025G BLDPBD29187-25G BLDPBD29187-5G BLDPBD29187-10G BLDPBD29187-100G
2-Acetyl-4-methylpyridine 98%
2-Acetyl-4-methylpyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₉NO
MW: 135,1632 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01863638
CAS-nummer: 59576-26-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR