4-Chloro-1-methylpiperidine 97%

BLDPBD2965-100GEA 26 EUR
BLDPBD2965-100G BLDPBD2965-10G BLDPBD2965-500G BLDPBD2965-25G
4-Chloro-1-methylpiperidine 97%
4-Chloro-1-methylpiperidine

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₁₂ClN
MW: 133,6192 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00044489
CAS-nummer: 5570-77-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR