6-Bromoquinazoline 96%

BLDPBD29851-25GEA 769 EUR
BLDPBD29851-25G BLDPBD29851-250MG BLDPBD29851-10G BLDPBD29851-1G BLDPBD29851-100MG BLDPBD29851-5G
6-Bromoquinazoline 96%
6-Bromoquinazoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₈H₅BrN₂
MW: 209,0400 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08236695
CAS-nummer: 89892-21-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR