Potassium (4-pyridyl)trifluoroborate 95%

BLDPBD306104-25GEA 500 EUR
BLDPBD306104-25G BLDPBD306104-5G BLDPBD306104-1G BLDPBD306104-250MG
Potassium (4-pyridyl)trifluoroborate 95%
Potassium (4-pyridyl)trifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Formule: C₅H₄BF₃KN
MW: 184,9965 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD09038543
CAS-nummer: 1111732-87-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR