4-Bromo-2,6-difluorophenol 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD3113-100GEA 306 EUR
BLDPBD3113-100G BLDPBD3113-10G BLDPBD3113-5G BLDPBD3113-25G BLDPBD3113-1G BLDPBD3113-500G
4-Bromo-2,6-difluorophenol 97%
4-Bromo-2,6-difluorophenol

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₃BrF₂O
MW: 208,99 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00236213
CAS-nummer: 104197-13-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR