4-(Cyclopropylamino)benzonitrile 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD316379-1GEA 37 EUR
BLDPBD316379-1G BLDPBD316379-5G BLDPBD316379-25G
4-(Cyclopropylamino)benzonitrile 98%
4-(Cyclopropylamino)benzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₀H₁₀N₂
MW: 158,20 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD11135501
CAS-nummer: 1019607-55-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR