2,3,6-Trifluorobenzonitrile 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD31830-250MGEA 15 EUR
BLDPBD31830-250MG BLDPBD31830-5G BLDPBD31830-25G BLDPBD31830-1G
2,3,6-Trifluorobenzonitrile 97%
2,3,6-Trifluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₂F₃N
MW: 157,09 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00012287
CAS-nummer: 136514-17-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR