6-Chloro-4-quinazolone 97%

BLDPBD32149-5GEA 16 EUR
BLDPBD32149-5G BLDPBD32149-25G BLDPBD32149-1G BLDPBD32149-250MG BLDPBD32149-100G BLDPBD32149-10G
6-Chloro-4-quinazolone 97%
6-Chloro-4-quinazolone

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₅ClN₂O
MW: 180,5900 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01686365
CAS-nummer: 16064-14-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR