Methyl-1H-imidazole-5-carboxylate 98%

BLDPBD32273-1GEA 7 EUR
BLDPBD32273-1G BLDPBD32273-100G BLDPBD32273-5G BLDPBD32273-10G BLDPBD32273-500G BLDPBD32273-25G
Methyl-1H-imidazole-5-carboxylate 98%
Methyl-1H-imidazole-5-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₅H₆N₂O₂
MW: 126,11 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00216584
CAS-nummer: 17325-26-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR