3-Bromo-4-chloroanisole 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD32987-5GEA 15 EUR
BLDPBD32987-5G BLDPBD32987-10G BLDPBD32987-1G
3-Bromo-4-chloroanisole 97%
3-Bromo-4-chloroanisole

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₆BrClO
MW: 221,48 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00070739
CAS-nummer: 2732-80-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR