3-Fluoro-2-methox-6-methylypyridine 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD33450-1GEA 80 EUR
BLDPBD33450-1G BLDPBD33450-250MG
3-Fluoro-2-methox-6-methylypyridine 97%
3-Fluoro-2-methox-6-methylypyridine
Formule: C₇H₈FNO
MW: 141,15 g/mol
MDL-nummer: MFCD03095083
CAS-nummer: 375368-80-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR