2-(3,5-Dimethoxyphenyl)propan-2-ol 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD33525-100MGEA 23 EUR
BLDPBD33525-100MG BLDPBD33525-5G BLDPBD33525-10G BLDPBD33525-250MG BLDPBD33525-1G
2-(3,5-Dimethoxyphenyl)propan-2-ol 97%
2-(3,5-Dimethoxyphenyl)propan-2-ol

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₁H₁₆O₃
MW: 196,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02684175
CAS-nummer: 39507-96-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR