Indoline 97%

BLDPBD33790-500GEA 76 EUR
BLDPBD33790-500G BLDPBD33790-0.5G BLDPBD33790-10G BLDPBD33790-25G BLDPBD33790-100G BLDPBD33790-5G BLDPBD33790-1000G
Indoline 97%
Indoline

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₉N
MW: 119,17 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005705
CAS-nummer: 496-15-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR