5-Bromo-2-chlorotoluene 98%

BLDPBD34056-5GEA 7 EUR
BLDPBD34056-5G BLDPBD34056-100G BLDPBD34056-25G BLDPBD34056-0.025G BLDPBD34056-500G BLDPBD34056-1G BLDPBD34056-10G
5-Bromo-2-chlorotoluene 98%
5-Bromo-2-chlorotoluene

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₆BrCl
MW: 205,48 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00018503
CAS-nummer: 54932-72-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR