5-Bromo-m-xylene 98%

BLDPBD34094-5GEA 7 EUR
BLDPBD34094-5G BLDPBD34094-500G BLDPBD34094-100G BLDPBD34094-25G BLDPBD34094-1000G BLDPBD34094-10G
5-Bromo-m-xylene 98%
5-Bromo-m-xylene

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₉Br
MW: 185,06 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00000087
CAS-nummer: 556-96-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR