L(+)-Glutamine 98%

BLDPBD34121-1GEA 0 EUR
BLDPBD34121-1G BLDPBD34121-100G BLDPBD34121-25G BLDPBD34121-500G BLDPBD34121-1000G
L(+)-Glutamine 98%
L(+)-Glutamine

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formule: C₅H₁₀N₂O₃
MW: 146,15 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD00008044
CAS-nummer: 56-85-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR