5-Aminovaleric acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD34544-1GEA 7 EUR
BLDPBD34544-1G BLDPBD34544-25G BLDPBD34544-100G BLDPBD34544-500G BLDPBD34544-10G BLDPBD34544-5G
5-Aminovaleric acid 98%
5-Aminovaleric acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₅H₁₁NO₂
MW: 117,15 g/mol
Opslagtemperatuur: Kamertemperatuur
CAS-nummer: 660-88-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR