(3-Bromobenzyl)methylamine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD34577-25GEA 84 EUR
BLDPBD34577-25G BLDPBD34577-10G BLDPBD34577-100G BLDPBD34577-5G BLDPBD34577-1G
(3-Bromobenzyl)methylamine 98%
(3-Bromobenzyl)methylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₈H₁₀BrN
MW: 200,08 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04507522
CAS-nummer: 67344-77-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR