2-Oxo-1,2,3,4-tetrahydroquinoline-6-carboxylic acid 97%

BLDPBD34696-1GEA 27 EUR
BLDPBD34696-1G BLDPBD34696-25G BLDPBD34696-250MG BLDPBD34696-5G
2-Oxo-1,2,3,4-tetrahydroquinoline-6-carboxylic acid 97%
2-Oxo-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₉NO₃
MW: 191,1834 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08271842
CAS-nummer: 70639-77-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR