2-Amino-4-fluorfenol 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD3499-10GEA 26 EUR
BLDPBD3499-10G BLDPBD3499-5G BLDPBD3499-25G BLDPBD3499-1G
2-Amino-4-fluorfenol 97%
2-Amino-4-fluorfenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₆FNO
MW: 127,12 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00077451
CAS-nummer: 399-97-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR