5,6-Difluoroindoline-2,3-dione 97%

BLDPBD35056-100GEA 971 EUR
BLDPBD35056-100G BLDPBD35056-250MG BLDPBD35056-5G BLDPBD35056-25G BLDPBD35056-1G
5,6-Difluoroindoline-2,3-dione 97%
5,6-Difluoroindoline-2,3-dione

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₃F₂NO₂
MW: 183,110 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02998377
CAS-nummer: 774-47-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR