α-Bromo-2,5-dichlorotoluene 98%

BLDPBD35135-5GEA 37 EUR
BLDPBD35135-5G BLDPBD35135-1G BLDPBD35135-25G BLDPBD35135-10G
α-Bromo-2,5-dichlorotoluene 98%
α-Bromo-2,5-dichlorotoluene

Storage condition: Inert atmosphere, 2-8°C

Formule: C₇H₅BrCl₂
MW: 239,93 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00236034
CAS-nummer: 85482-13-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR