(±)-Boc-α-phosphonoglycine trimethyl ester 98%

BLDPBD35348-25GEA 89 EUR
BLDPBD35348-25G BLDPBD35348-100G BLDPBD35348-500G BLDPBD35348-1G BLDPBD35348-0.025G BLDPBD35348-5G BLDPBD35348-10G
(±)-Boc-α-phosphonoglycine trimethyl ester 98%
(±)-Boc-α-phosphonoglycine trimethyl ester

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₁₀H₂₀NO₇P
MW: 297,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD02178893
CAS-nummer: 89524-98-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR