Nicotinamidine hydrochloride 95%

Synoniemen: Pyridine-3-carboximidamide MonohydrochloridePyridine-3-carboximidamide hydrochloride3-Amidinopyridine hydrochloride3-Carbamimidoylpyridine Monohydrochloride3-Amidinopyridinium chloride

BLDPBD3621-5GEA 18 EUR
BLDPBD3621-5G BLDPBD3621-100G BLDPBD3621-1G BLDPBD3621-25G
Nicotinamidine hydrochloride 95%
Nicotinamidine hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₈ClN₃
MW: 157,60 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00054326
CAS-nummer: 7356-60-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR