2-Fluoro-4-iodonicotinic acid 95%

BLDPBD37098-10GEA 151 EUR
BLDPBD37098-10G BLDPBD37098-1G BLDPBD37098-25G BLDPBD37098-250MG BLDPBD37098-5G
2-Fluoro-4-iodonicotinic acid 95%
2-Fluoro-4-iodonicotinic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₃FINO₂
MW: 267,0000 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03095310
CAS-nummer: 884494-51-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR